Potassium hexacyanoferrate(III), Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

455946-100G 455946-25G
SIAL455946-100GEA 1040 EUR
SIAL455946-100G SIAL455946-25G
Potassium hexacyanoferrate(III), Sigma-Aldrich®
Potassium hexacyanoferrate (III)
Formula: K₃Fe(CN)₆
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR