Potassium iodide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dipotassium diiodide, Kali iodide
MQ200
204102-10G 204102-250G 204102-50G
SIAL204102-10GEA 68.8 EUR
SIAL204102-10G SIAL204102-250G SIAL204102-50G
Potassium iodide, Sigma-Aldrich®
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727