Sodium iodide
Supplier: Roth Carl
Warning
8783.1 8783.2
ROTH8783.1EA 103 EUR
ROTH8783.1 ROTH8783.2
Sodium iodide
Sodium iodide
Formula: NaI
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 662 °C
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705