Cesium nitrate ≥99.99% (trace metals basis), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nitric acid cesium salt
M-Clarity™ quality level = MQ100
202150-10G 202150-50G
SIAL202150-10GEA 45 EUR
SIAL202150-10G SIAL202150-50G
Cesium nitrate ≥99.99% (trace metals basis), Sigma-Aldrich®
Cesium nitrate
Formula: CsNO₃
Boiling Pt: 737 °C (1013 hPa)
Melting Pt: 414 °C
Density: 3.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010963
CAS Number: 7789-18-6
EINECS: 232-146-8
UN: 1451
ADR: 5.1,III
Merck Index: 12,02057