Potassium nitrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Nitrate of Potash
M-Clarity™ quality level = MQ100
P6083-1KG P6083-500G
SIALP6083-1KGEA 263 EUR
SIALP6083-1KG SIALP6083-500G
Potassium nitrate, Sigma-Aldrich®
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733