Rubidium nitrate, Sigma-Aldrich®

Supplier: Merck
Danger

204269-10G 204269-1G
SIAL204269-10GEA 301 EUR
SIAL204269-10G SIAL204269-1G
Rubidium nitrate, Sigma-Aldrich®
Rubidium nitrate
Formula: NO₃Rb
Boiling Pt: 578 °C (1013 hPa)
Melting Pt: 310 °C
Density: 3.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011193
CAS Number: 13126-12-0
EINECS: 236-060-1
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR