Dilithium oxide, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Lithium oxide

M-Clarity™ quality level = MQ100

374725-100G 374725-10G
SIAL374725-100GEA 552 EUR
SIAL374725-100G SIAL374725-10G
Dilithium oxide, Sigma-Aldrich®
di-Lithium oxide
Formula: Li₂O
MW: 29.88 g/mol
Melting Pt: 1450 °C
Density: 2.013 g/cm³ (25 °C)
MDL Number: MFCD00016183
CAS Number: 12057-24-8
EINECS: 235-019-5
UN: 3262
ADR: 8,II
Merck Index: 13,05560

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR