Potassium hydroxide
Supplier: Cayman Chemical
Danger
400029-1 400029-5
CAYM400029-1EA 16 EUR
CAYM400029-1 CAYM400029-5
Potassium hydroxide
Potassium hydroxide
Formula: KOH
Boiling Pt: 1320 °C (1013 hPa)
Melting Pt: 360 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
REACH: 01-2119487136-33
Merck Index: 12,07806