Potassium hexafluorophosphate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
200913-1KG 200913-250G 200913-50G
SIAL200913-1KGEA 439 EUR
SIAL200913-1KG SIAL200913-250G SIAL200913-50G
Potassium hexafluorophosphate, Sigma-Aldrich®
Potassium hexafluorophosphate
Formula: F₆KP
MW: 184.06 g/mol
Melting Pt: 575 °C
Density: 2.58 g/cm³ (20 °C)
MDL Number: MFCD00011412
CAS Number: 17084-13-8
EINECS: 241-143-0
UN: 3260
ADR: 8,II