Tripotassium orthophosphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Potassium phosphate tribasic, Potassium phosphate
M-Clarity™ quality level = MQ200
P5629-25G P5629-500G P5629-10KG
SIALP5629-25GEA 32.9 EUR
SIALP5629-25G SIALP5629-500G SIALP5629-10KG
Tripotassium orthophosphate, Sigma-Aldrich®
tri-Potassium orthophosphate
Formula: K₃O₄P
MW: 212.27 g/mol
Melting Pt: 1340 °C
Density: 2.564 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036295
CAS Number: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830