Sodium phosphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: tri-Sodium phosphate, Sodium phosphate tribasic, Trisodium phosphate
M-Clarity™ quality level = MQ200
342483-2.5KG 342483-25G 342483-500G
SIAL342483-2.5KGEA 156 EUR
SIAL342483-2.5KG SIAL342483-25G SIAL342483-500G
Sodium phosphate, Sigma-Aldrich®
tri-Sodium phosphate
Formula: Na₃O₄P
MW: 163.94 g/mol
Melting Pt: 73.3…76.7 °C
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003510
CAS Number: 7601-54-9
EINECS: 231-509-8
UN: 3262
ADR: 8 III
Merck Index: 12,08808