Sodium dihydrogen phosphate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic

MQ300

S8282-1KG S8282-500G
SIALS8282-1KGEA 458 EUR
SIALS8282-1KG SIALS8282-500G
Sodium dihydrogen phosphate, Sigma-Aldrich®
Sodium dihydrogen phosphate
Formula: NaH₂PO₄
MW: 119.98 g/mol
Melting Pt: ∼650 °C
Density: 0.9996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR