Potassium tetrachloroplatinate(II) Pt 46-47%
Supplier: Thermo Scientific
Danger
Synonyms: Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate
195360010 195360050 195360250
ACRO195360010EA 136 EUR
ACRO195360010 ACRO195360050 ACRO195360250
Potassium tetrachloroplatinate(II) Pt 46-47%
Potassium tetrachloroplatinate (II)
Formula: Cl₄K₂Pt
MW: 415.09 g/mol
Boiling Pt: 35…40 °C (20 mmHg)
Melting Pt: 250 °C (dec.)
Density: 3,38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III
Merck Index: 12,07855
Order Now
Specification Test Results
Appearance Red-brown Crystalline powder or crystals
Total trace metal impurities ≤500 ppm (referring to Pt)
Gravimetry 46 to 47 % (Pt)