Potassium tetrachloroplatinate(II), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate
M-Clarity™ quality level = MQ100
323411-1G 323411-250MG 323411-5G
SIAL323411-1GEA 248 EUR
SIAL323411-1G SIAL323411-250MG SIAL323411-5G
Potassium tetrachloroplatinate(II), Sigma-Aldrich®
Potassium tetrachloroplatinate (II)
Formula: Cl₄K₂Pt
MW: 415.09 g/mol
Boiling Pt: 35…40 °C (20 mmHg)
Melting Pt: 265 °C
Density: 3.38 g/cm³ (20 °C)
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III
Merck Index: 12,07855