Disodium sulphide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Sodium sulphide
M-Clarity™ quality level = MQ200
407410-10G
407410-50G
SIAL407410-10GEA
182
EUR
SIAL407410-10G
SIAL407410-50G
Disodium sulphide, Sigma-Aldrich®
Disodium sulphide
Formula:
Na₂S MW: 78.05 g/mol Boiling Pt: 174 °C (1013 hPa) Density: 1.856 g/cm³ (14 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003498 CAS Number: 1313-82-2 EINECS: 215-211-5 UN: 1385 ADR: 4.2,II REACH: 01-2119513694-38 Merck Index: 12,08830 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...