Sodium thiosulphate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
M-Clarity™ quality level = MQ100
563188-100G 563188-25G
SIAL563188-100GEA 442 EUR
SIAL563188-100G SIAL563188-25G
Sodium thiosulphate, Sigma-Aldrich®
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844