Potassium peroxodisulphate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: di-Potassium peroxodisulphate, Peroxydisulphuric acid dipotassium salt, Dipotassium peroxodisulphate, Potassium persulphate

M-Clarity™ quality level = MQ200

379824-25G 379824-5G
SIAL379824-25GEA 288 EUR
SIAL379824-25G SIAL379824-5G
Potassium peroxodisulphate, Sigma-Aldrich®
Potassium peroxodisulphate
Formula: K₂S₂O₈
Melting Pt: 100 °C
Density: 2.477 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III
Merck Index: 13,07741

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR