Sodium selenite, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIALS9133-1MGEA 123 EUR
SIALS9133-1MG
Sodium selenite, Sigma-Aldrich®
Sodium selenite
Formula: Na₂SeO₃
MW: 172,94 g/mol
Melting Pt: 710 °C
Density: 3,1 g/cm³ (20 °C)
MDL Number: MFCD00003489
UN: 2630
ADR: 6.1,I
Merck Index: 12,08822