Sodium sulphite, Sigma-Aldrich®
Supplier: Merck
MQ200
S4672-100G S4672-1KG S4672-250G
SIALS4672-100GEA 75.5 EUR
SIALS4672-100G SIALS4672-1KG SIALS4672-250G
Sodium sulphite, Sigma-Aldrich®
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: > 500 °C
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757