Cesium sulphate 99
Supplier: Thermo Fisher Scientific
Warning
A16767.14 A16767.22 A16767.36
A16767.14EA 39 EUR
A16767.14 A16767.22 A16767.36
Cesium sulphate 99
Cesium sulphate
Purity: 99% CAS No.: 10294-54-9 Molecular Formula: Cs2SO4 Formula Weight: 361.87 Odour: Odourless Melting Point: 1010° MDL No.: MFCD00010959
Formula: Cs₂O₄S
MW: 361.87 g/mol
Boiling Pt: 1627
Melting Pt: 1010
Density: 4.24
Storage Temperature: Ambient
MDL Number: MFCD00010959
CAS Number: 10294-54-9
EINECS: 233-662-6
Merck Index: 13,02025