Potassium sulphate
Supplier: Roth Carl
X889.1 X889.2 X889.3 X889.5
ROTHX889.1EA 19.1 EUR
ROTHX889.1 ROTHX889.2 ROTHX889.3 ROTHX889.5
Potassium sulphate
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759