Potassium sulphate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
204129-10G 204129-50G
SIAL204129-10GEA 127 EUR
SIAL204129-10G SIAL204129-50G
Potassium sulphate, Sigma-Aldrich®
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759