Potassium sulphate, Sigma-Aldrich®
Supplier: Merck
60528-1KG 60528-250G
SIAL60528-1KGEA 267 EUR
SIAL60528-1KG SIAL60528-250G
Potassium sulphate, Sigma-Aldrich®
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759