Potassium hydrogen sulphate
Supplier: Molekula
MOLE19700986-1KGEA 109 EUR
MOLE19700986-1KG
Potassium hydrogen sulphate
Potassium hydrogen sulphate
Formula: KHSO₄
MW: 136,17 g/mol
Melting Pt: 214 °C
Density: 2,32 g/cm³ (20 °C)
MDL Number: MFCD00011404
UN: 2509
ADR: 8,II
Merck Index: 13,07695