Sodium orthovanadate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
S6508-10G S6508-250G S6508-50G
SIALS6508-10GEA 61.5 EUR
SIALS6508-10G SIALS6508-250G SIALS6508-50G
Sodium orthovanadate, Sigma-Aldrich®
Sodium orthovanadate
Formula: Na₃O₄V
MW: 183.91 g/mol
Melting Pt: 850…866 °C
Storage Temperature: Ambient
MDL Number: MFCD00003511
CAS Number: 13721-39-6
EINECS: 237-287-9