Calcium bromide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

413631-1G 413631-5G
SIAL413631-1GEA 43.5 EUR
SIAL413631-1G SIAL413631-5G
Calcium bromide, Sigma-Aldrich®
Calcium bromide
Formula: Br₂Ca
MW: 199.89 g/mol
Boiling Pt: 810 °C (1013 hPa)
Melting Pt: ∼730 °C
Density: 3.353 g/cm³ (20 °C)
MDL Number: MFCD00010902
CAS Number: 7789-41-5
EINECS: 232-164-6
Merck Index: 12,01693

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR