Magnesium bromide, Sigma-Aldrich®

Supplier: Merck
Warning


MQ200

360074-10G 360074-50G
SIAL360074-10GEA 43.5 EUR
SIAL360074-10G SIAL360074-50G
Magnesium bromide, Sigma-Aldrich®
Magnesium bromide
Formula: Br₂Mg
MW: 184.11 g/mol
Melting Pt: 700
Density: 3.72 g/cm³ (20 °C)
MDL Number: MFCD00011105
CAS Number: 7789-48-2
EINECS: 232-170-9
Merck Index: 12,05695

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR