Magnesium bromide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
MQ100
495093-1G
495093-5G
SIAL495093-1GEA
154
EUR
SIAL495093-1G
SIAL495093-5G
Magnesium bromide, Sigma-Aldrich®
Magnesium bromide
Formula:
Br₂Mg MW: 184.11 g/mol Melting Pt: 700 Density: 3.72 g/cm³ (20 °C) |
MDL Number:
MFCD00011105 CAS Number: 7789-48-2 EINECS: 232-170-9 Merck Index: 12,05695 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...