Barium chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIAL202738-5GEA
48.7
EUR
SIAL202738-5G
SIAL202738-25G
SIAL202738-100G
Barium chloride, Sigma-Aldrich®
Barium chloride
Formula:
BaCl₂ MW: 208,24 g/mol Boiling Pt: 1560 °C (1013 hPa) Melting Pt: ∼963 °C Density: 2,33 g/cm³ (20 °C) |
MDL Number:
MFCD00003445 UN: 1564 ADR: 6.1,III Merck Index: 12,00998 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...