Barium chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIAL342920-250GEA 433 EUR
SIAL342920-250G SIAL342920-50G SIAL342920-10G
Barium chloride, Sigma-Aldrich®
Barium chloride
Formula: BaCl₂
MW: 208,24 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: ∼963 °C
Density: 2,33 g/cm³ (20 °C)
MDL Number: MFCD00003445
UN: 1564
ADR: 6.1,III
Merck Index: 12,00998