Calcium chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
M-Clarity™ quality level = MQ200
C5670-100G C5670-500G
SIALC5670-100GEA 158 EUR
SIALC5670-100G SIALC5670-500G
Calcium chloride, Sigma-Aldrich®
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659