Magnesium chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
M8266-100G M8266-1KG
SIALM8266-100GEA 59.4 EUR
SIALM8266-100G SIALM8266-1KG
Magnesium chloride, Sigma-Aldrich®
Magnesium chloride
Formula: MgCl₂
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 715 °C
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684