Calcium carbonate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Marble
MQ100
310034-100G
310034-500G
SIAL310034-100GEA
38.8
EUR
SIAL310034-100G
SIAL310034-500G
Calcium carbonate, Sigma-Aldrich®
Calcium carbonate
Formula:
CaCO₃ MW: 100.09 g/mol Melting Pt: 825 Density: 2.71 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...