Calcium carbonate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Marble

MQ200

398101-100G 398101-500G
SIAL398101-100GEA 34.9 EUR
SIAL398101-100G SIAL398101-500G
Calcium carbonate, Sigma-Aldrich®
Calcium carbonate
Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825
Density: 2.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR