Calcium carbonate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Marble
MQ200
C4830-100G C4830-500G
SIALC4830-100GEA 71.5 EUR
SIALC4830-100G SIALC4830-500G
Calcium carbonate, Sigma-Aldrich®
Calcium carbonate
Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825
Density: 2.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658