Magnesium carbonate heavy heavy, extra pure
Supplier: Thermo Scientific
Synonyms: Magnesium carbonate hydroxide, Magnesium carbonate basic, heavy
413405000 413400025 413400100
ACRO413405000EA 43.9 EUR
ACRO413405000 ACRO413400025 ACRO413400100
Magnesium carbonate heavy heavy, extra pure
Magnesium carbonate heavy
Formula: CH₂Mg₂O₅
MW: 142.63 g/mol
Density: 3,03 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011107
CAS Number: 39409-82-0
EINECS: 208-915-9
Order Now
Specification Test Results
Appearance White Powder
Titration Complexometric 40 to 45 % (MgO)
Loss on calcination 55 % to 60 %
Titration with HClO4 ≥96.0 %