Strontium carbonate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
289833-1KG 289833-250G
SIAL289833-1KGEA 74.8 EUR
SIAL289833-1KG SIAL289833-250G
Strontium carbonate, Sigma-Aldrich®
Strontium carbonate
Formula: CO₃Sr
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1768.85 °C
Density: 3.548 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,08838