Barium fluoride, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
652458-100G 652458-25G
SIAL652458-100GEA 270 EUR
SIAL652458-100G SIAL652458-25G
Barium fluoride, Sigma-Aldrich®
Barium fluoride
Formula: BaF₂
MW: 175.33 g/mol
Boiling Pt: 2260
Melting Pt: 1270
Density: 4.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1 III
Merck Index: 12,01003