Magnesium iodide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

394599-25G 394599-5G
SIAL394599-25GEA 284 EUR
SIAL394599-25G SIAL394599-5G
Magnesium iodide, Sigma-Aldrich®
Magnesium iodide
Formula: MgI₂
MW: 278.11 g/mol
Melting Pt: 637
Density: 3.89 g/cm³ (20 °C)
MDL Number: MFCD00049485
CAS Number: 10377-58-9
EINECS: 233-825-1
Merck Index: 12,05707

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR