Strontium nitrate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
204498-10G
204498-50G
SIAL204498-10GEA
133
EUR
SIAL204498-10G
SIAL204498-50G
Strontium nitrate, Sigma-Aldrich®
Strontium nitrate
Formula:
Sr(NO₃)₂ MW: 211.63 g/mol Boiling Pt: 645 °C (1013 hPa) Melting Pt: 570 °C Density: 2.113 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011248 CAS Number: 10042-76-9 EINECS: 233-131-9 UN: 1507 ADR: 5.1,III Merck Index: 13,08926 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...