Barium oxide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
288497-250G 288497-50G
SIAL288497-250GEA 889 EUR
SIAL288497-250G SIAL288497-50G
Barium oxide, Sigma-Aldrich®
Barium oxide
Formula: BaO
MW: 153.33 g/mol
Boiling Pt: ∼2000 °C (1013 hPa)
Melting Pt: ∼1920 °C
Density: ∼5.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003453
CAS Number: 1304-28-5
EINECS: 215-127-9
UN: 1884
ADR: 6.1 III
Merck Index: 12,01015