Calcium oxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime
M-Clarity™ quality level = MQ100
208159-100G 208159-25G 208159-500G
SIAL208159-100GEA 188 EUR
SIAL208159-100G SIAL208159-25G SIAL208159-500G
Calcium oxide, Sigma-Aldrich®
Calcium oxide
Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850 °C (1013 hPa)
Melting Pt: 2580 °C
Density: 3.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
Merck Index: 13,01691