Magnesium oxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
529699-10G 529699-50G
SIAL529699-10GEA 148 EUR
SIAL529699-10G SIAL529699-50G
Magnesium oxide, Sigma-Aldrich®
Magnesium oxide
Formula: MgO
Boiling Pt: 3600 °C (1013 hPa)
Melting Pt: 2800 °C
Density: 3.58 g/cm³ (25 °C)
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700