Calcium hydroxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Slaked lime, Calcium dihydroxide
MQ100
450146-25G 450146-5G
SIAL450146-25GEA 295 EUR
SIAL450146-25G SIAL450146-5G
Calcium hydroxide, Sigma-Aldrich®
Calcium hydroxide
Formula: Ca(OH)₂
MW: 74.09 g/mol
Melting Pt: 550
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8 III
Merck Index: 13,01675