Magnesium hydroxide 95%, pure
Supplier: Thermo Scientific
Warning
315725000 315720025 315720250
ACRO315725000EA 46.1 EUR
ACRO315725000 ACRO315720025 ACRO315720250
Magnesium hydroxide 95%, pure
Magnesium hydroxide
Formula: Mg(OH)₂
MW: 58.32 g/mol
Melting Pt: 350 °C (dec.)
Density: 2,3261 to 2.3631 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693
Order Now
Specification Test Results
Appearance White Powder
Titration Complexometric ≥94 %