Aluminium bromide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Aluminum bromide

M-Clarity™ quality level = MQ100

210072-100G 210072-25G
SIAL210072-100GEA 165 EUR
SIAL210072-100G SIAL210072-25G
Aluminium bromide, Sigma-Aldrich®
Aluminium bromide
Formula: AlBr₃
MW: 266.69 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 97 °C
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003421
CAS Number: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8,II
Merck Index: 13,00330

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR