Boron tribromide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
230367-25G 230367-5G
SIAL230367-25GEA 94.7 EUR
SIAL230367-25G SIAL230367-5G
Boron tribromide, Sigma-Aldrich®
Boron tribromide
Formula: BBr₃
MW: 250.52 g/mol
Boiling Pt: 91.2 °C (1013 hPa)
Melting Pt: –46 °C
Density: 2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8,I
Merck Index: 13,01337