Thallium(I) bromide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Thallium bromide
M-Clarity™ quality level = MQ100
336270-10G 336270-50G
SIAL336270-10GEA 125 EUR
SIAL336270-10G SIAL336270-50G
Thallium(I) bromide, Sigma-Aldrich®
Thallium (I) bromide
Formula: BrTl
MW: 284.29 g/mol
Boiling Pt: 815 °C (1013 hPa)
Melting Pt: 460 °C
Density: 7.56 g/cm³ (20 °C)
MDL Number: MFCD00011273
CAS Number: 7789-40-4
EINECS: 232-163-0
UN: 1707
ADR: 6.1,II
Merck Index: 12,09393