Aluminium chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Aluminum chloride
MQ100
563919-25G 563919-5G
SIAL563919-25GEA 378 EUR
SIAL563919-25G SIAL563919-5G
Aluminium chloride, Sigma-Aldrich®
Aluminium chloride
Formula: AlCl₃
MW: 133.34 g/mol
Boiling Pt: 180
Melting Pt: 180…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8 II
Merck Index: 13,00335