Indium(III) chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Indium Trichloride Chloride, Indium trichloride
M-Clarity™ quality level = MQ200
429414-25G 429414-5G
SIAL429414-25GEA 505 EUR
SIAL429414-25G SIAL429414-5G
Indium(III) chloride, Sigma-Aldrich®
Indium (III) chloride
Formula: Cl₃In
MW: 221.18 g/mol
Boiling Pt: 800
Melting Pt: 586
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011058
CAS Number: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8 II
Merck Index: 13,04980